8-Amino-3-(3,4-dichlorphenyl)imidazo[1,2-a]pyridin

Lieferant: BIONET RESEARCH

3M-917.500 3M-917.1 3M-917.5 3M-917.10
BIOT3M-917.500EA 0 EUR
BIOT3M-917.500 BIOT3M-917.1 BIOT3M-917.5 BIOT3M-917.10
8-Amino-3-(3,4-dichlorphenyl)imidazo[1,2-a]pyridin
8-Amino-3-(3,4-dichlorphenyl)imidazo[1,2-a]pyridin
Formel: C₁₃H₉Cl₂N₃
MDL: MFCD00975027
CAS-Nummer: 338415-23-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR