8-Amino-3-(4-chlorphenyl)imidazo[1,2-a]pyridin

Lieferant: BIONET RESEARCH

Synonyme: 3-(4-Chlorophenyl)imidazo[1,2-a]pyridin-8-amine

3M-903.500 3M-903.1 3M-903.5 3M-903.10
BIOT3M-903.500EA 0 EUR
BIOT3M-903.500 BIOT3M-903.1 BIOT3M-903.5 BIOT3M-903.10
8-Amino-3-(4-chlorphenyl)imidazo[1,2-a]pyridin
8-Amino-3-(4-chlorphenyl)imidazo[1,2-a]pyridin
Formel: C₁₃H₁₀ClN₃
MDL: MFCD00975015
CAS-Nummer: 338415-10-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR