2-(4-Fluorphenyl)imidazo[1,2-a]pyridin

Lieferant: COMBI-BLOCKS

HC-2084-5G HC-2084-1G
COBBHC-2084-5GEA 1180 EUR
COBBHC-2084-5G COBBHC-2084-1G
2-(4-Fluorphenyl)imidazo[1,2-a]pyridin
2-(4-Fluorphenyl)imidazo[1,2-a]pyridin
Formel: C₁₃H₉FN₂
Molecular Weight: 212,220 g/mol
MDL: MFCD00269178
CAS-Nummer: 347-12-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR