3,6-Di(imidazol-1-yl)pyridazin

Lieferant: COMBI-BLOCKS

Synonyme: 3,6-Di(1H-imidazol-1-yl)pyridazine

HC-6058-5G HC-6058-25G HC-6058-1G
COBBHC-6058-5GEA 451 EUR
COBBHC-6058-5G COBBHC-6058-25G COBBHC-6058-1G
3,6-Di(imidazol-1-yl)pyridazin
3,6-Di(imidazol-1-yl)pyridazin
Formel: C₁₀H₈N₆
Molecular Weight: 212,210 g/mol
MDL: MFCD09475834
CAS-Nummer: 177648-99-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR