2-(1,3-Dioxolan-2-yl)ethylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(2-(1,3-Dioxolan-2-yl)ethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-8611-1G PN-8611-250M
COBBPN-8611-1GEA 497 EUR
COBBPN-8611-1G COBBPN-8611-250M
2-(1,3-Dioxolan-2-yl)ethylboronsäurepinakolester
2-(1,3-Dioxolan-2-yl)ethylboronsäurepinakolester
Formel: C₁₁H₂₁BO₄
Molecular Weight: 228,10 g/mol
MDL: MFCD03788722
CAS-Nummer: 1073354-07-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR