(1H-Pyrazol-1-yl)essigsäure

Lieferant: COMBI-BLOCKS

Synonyme: 1-Pyrazoleacetic Acid

HI-1380-5G HI-1380-1G
COBBHI-1380-5GEA 0 EUR
COBBHI-1380-5G COBBHI-1380-1G
(1H-Pyrazol-1-yl)essigsäure
(1H-Pyrazol-1-yl)essigsäure
Formel: C₅H₆N₂O₂
MDL: MFCD00228774
CAS-Nummer: 16034-48-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR