1-(6-Chlor-3-pyridazinyl)-4-piperidincarbonsäureamid

Lieferant: BIONET RESEARCH

Synonyme: 1-(6-Chloro-3-pyridazinyl)piperidine-4-carboxamide

11F-905.500 11F-905.1 11F-905.5 11F-905.10
BIOT11F-905.500EA 0 EUR
BIOT11F-905.500 BIOT11F-905.1 BIOT11F-905.5 BIOT11F-905.10
1-(6-Chlor-3-pyridazinyl)-4-piperidincarbonsäureamid
1-(6-Chlor-3-pyridazinyl)-4-piperidincarbonsäureamid
Formel: C₁₀H₁₃ClN₄O
Molecular Weight: 240.69 g/mol
MDL: MFCD00180782
CAS-Nummer: 303149-97-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR