1-(6-Chlor-3-pyridazinyl)-4-piperidincarbonsäurehydrazid

Lieferant: BIONET RESEARCH

3L-905.500 3L-905.1 3L-905.5 3L-905.10
BIOT3L-905.500EA 0 EUR
BIOT3L-905.500 BIOT3L-905.1 BIOT3L-905.5 BIOT3L-905.10
1-(6-Chlor-3-pyridazinyl)-4-piperidincarbonsäurehydrazid
1-(6-Chlor-3-pyridazinyl)-4-piperidincarbonsäurehydrazid
Formel: C₁₀H₁₄ClN₅O
MDL: MFCD00794640
CAS-Nummer: 338413-80-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR