1-(6-Chlor-3-pyridazinyl)-4-piperidincarbonsäure

Lieferant: BIONET RESEARCH

4J-920.500 4J-920.1 4J-920.5 4J-920.10
BIOT4J-920.500EA 0 EUR
BIOT4J-920.500 BIOT4J-920.1 BIOT4J-920.5 BIOT4J-920.10
1-(6-Chlor-3-pyridazinyl)-4-piperidincarbonsäure
1-(6-Chlor-3-pyridazinyl)-4-piperidincarbonsäure
Formel: C₁₀H₁₂ClN₃O₂
MDL: MFCD00664523
CAS-Nummer: 339276-36-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR