Ethyl-6,7-dichlor-3-(trifluormethyl)-1,4-dihydrochinoxalin-2-carboxylat
Lieferant: COMBI-BLOCKS
CS-9168-250M
CS-9168-100M
COBBCS-9168-250MEA
1180
EUR
COBBCS-9168-250M
COBBCS-9168-100M
Ethyl-6,7-dichlor-3-(trifluormethyl)-1,4-dihydrochinoxalin-2-carboxylat
Ethyl-6,7-dichlor-3-(trifluormethyl)-1,4-dihydrochinoxalin-2-carboxylat
Formel:
C₁₂H₉Cl₂F₃N₂O₂ |
MDL:
MFCD09878367 CAS-Nummer: 957062-92-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...