4-(4-Brom-1H-pyrazol-1-yl)-N-(furan-2-ylmethyl)benzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-1110-5G OT-1110-1G
COBBOT-1110-5GEA 451 EUR
COBBOT-1110-5G COBBOT-1110-1G
4-(4-Brom-1H-pyrazol-1-yl)-N-(furan-2-ylmethyl)benzolsulfonamid
4-(4-Brom-1H-pyrazol-1-yl)-N-(furan-2-ylmethyl)benzolsulfonamid
Formel: C₁₄H₁₂BrN₃O₃S
MDL: MFCD13195743
CAS-Nummer: 1199773-46-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR