4-(4-Methylpiperazine-1-carbonyl)phenylboronsäure Hydrochlorid

Lieferant: COMBI-BLOCKS

Synonyme: 4-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid hydrochloride

BB-3213-5G BB-3213-1G
COBBBB-3213-5GEA 1590 EUR
COBBBB-3213-5G COBBBB-3213-1G
4-(4-Methylpiperazine-1-carbonyl)phenylboronsäure Hydrochlorid
4-(4-Methylpiperazine-1-carbonyl)phenylboronsäure Hydrochlorid
Formel: C₁₂H₁₈BClN₂O₃
Molecular Weight: 284,55 g/mol
MDL: MFCD09027217
CAS-Nummer: 913835-43-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR