3-Brom-N-(furan-2-ylmethyl)benzamid

Lieferant: COMBI-BLOCKS

CA-4477-5G CA-4477-1G
COBBCA-4477-5GEA 451 EUR
COBBCA-4477-5G COBBCA-4477-1G
3-Brom-N-(furan-2-ylmethyl)benzamid
3-Brom-N-(furan-2-ylmethyl)benzamid
Formel: C₁₂H₁₀BrNO₂
MDL: MFCD00504015
CAS-Nummer: 346663-79-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR