4-Brom-N-(furan-2-ylmethyl)benzamid
Lieferant: COMBI-BLOCKS
CA-4414-5G
CA-4414-25G
CA-4414-1G
COBBCA-4414-5GEA
451
EUR
COBBCA-4414-5G
COBBCA-4414-25G
COBBCA-4414-1G
4-Brom-N-(furan-2-ylmethyl)benzamid
4-Brom-N-(furan-2-ylmethyl)benzamid
Formel:
C₁₂H₁₀BrNO₂ |
MDL:
MFCD00451751 CAS-Nummer: 312587-75-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...