N-(Furan-2-ylmethyl)-2-brom-5-fluorbenzamid

Lieferant: COMBI-BLOCKS

CA-4440-5G CA-4440-1G
COBBCA-4440-5GEA 796 EUR
COBBCA-4440-5G COBBCA-4440-1G
N-(Furan-2-ylmethyl)-2-brom-5-fluorbenzamid
N-(Furan-2-ylmethyl)-2-brom-5-fluorbenzamid
Formel: C₁₂H₉BrFNO₂
MDL: MFCD08872270
CAS-Nummer: 923722-86-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR