2-Brom-N-(furan-2-ylmethyl)benzolsulfonamid
Lieferant: COMBI-BLOCKS
HC-6131-1G
HC-6131-5G
COBBHC-6131-1GEA
149
EUR
COBBHC-6131-1G
COBBHC-6131-5G
2-Brom-N-(furan-2-ylmethyl)benzolsulfonamid
2-Brom-N-(furan-2-ylmethyl)benzolsulfonamid
Formel:
C₁₁H₁₀BrNO₃S |
MDL:
MFCD07710748 CAS-Nummer: 849056-66-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...