4-Fluor-N-(furan-2-ylmethyl)benzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0956-5G OT-0956-1G
COBBOT-0956-5GEA 243 EUR
COBBOT-0956-5G COBBOT-0956-1G
4-Fluor-N-(furan-2-ylmethyl)benzolsulfonamid
4-Fluor-N-(furan-2-ylmethyl)benzolsulfonamid
Formel: C₁₁H₁₀FNO₃S
MDL: MFCD01077680
CAS-Nummer: 6341-35-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR