4-(4-Brom-1H-pyrazol-1-yl)benzolsulfonamid

Lieferant: COMBI-BLOCKS

HI-1316-5G HI-1316-1G
COBBHI-1316-5GEA 983 EUR
COBBHI-1316-5G COBBHI-1316-1G
4-(4-Brom-1H-pyrazol-1-yl)benzolsulfonamid
4-(4-Brom-1H-pyrazol-1-yl)benzolsulfonamid
Formel: C₉H₈BrN₃O₂S
MDL: MFCD09475848
CAS-Nummer: 957034-91-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR