4-(4-Brom-1H-pyrazol-1-yl)-N-tert-butylbenzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-1111-5G OT-1111-1G
COBBOT-1111-5GEA 303 EUR
COBBOT-1111-5G COBBOT-1111-1G
4-(4-Brom-1H-pyrazol-1-yl)-N-tert-butylbenzolsulfonamid
4-(4-Brom-1H-pyrazol-1-yl)-N-tert-butylbenzolsulfonamid
Formel: C₁₃H₁₆BrN₃O₂S
MDL: MFCD13195744
CAS-Nummer: 1199773-25-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR