4-(4-Brom-1H-pyrazol-1-yl)-N-isopropylbenzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0778-1G OT-0778-5G
COBBOT-0778-1GEA 98.3 EUR
COBBOT-0778-1G COBBOT-0778-5G
4-(4-Brom-1H-pyrazol-1-yl)-N-isopropylbenzolsulfonamid
4-(4-Brom-1H-pyrazol-1-yl)-N-isopropylbenzolsulfonamid
Formel: C₁₂H₁₄BrN₃O₂S
MDL: MFCD12756457
CAS-Nummer: 1187385-75-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR