4-(1H-Pyrazol-1-yl)benzolsulfonamid

Lieferant: COMBI-BLOCKS

HI-1290-1G HI-1290-5G
COBBHI-1290-1GEA 303 EUR
COBBHI-1290-1G COBBHI-1290-5G
4-(1H-Pyrazol-1-yl)benzolsulfonamid
4-(1H-Pyrazol-1-yl)benzolsulfonamid
Formel: C₉H₉N₃O₂S
MDL: MFCD11504936
CAS-Nummer: 51891-85-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR