2,3-Diphenylchinoxalin ≥98%

Lieferant: Thermo Fisher Scientific

B20100.06 B20100.14 B20100.22
B20100.06EA 32.4 EUR
B20100.06 B20100.14 B20100.22
2,3-Diphenylchinoxalin ≥98%
2,3-Diphenylquinoxaline

Purity: 98+% CAS No.: 1684-14-6 Molecular Formula: C20H14N2 Formula Weight: 282.35 Melting Point: 124-128° Beilstein Reference: 209272

Formel: C₂₀H₁₄N₂
Molecular Weight: 282.35 g/mol
Schmelzpunkt: 124…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014659
CAS-Nummer: 1684-14-6
EINECS: 216-865-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR