Tropyliumtetrafluoroborat ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Tropyliumtetrafluoroborate, Cyclohepta-2,4,6-trienylium tetrafluoroborate

A14045.06 A14045.14 A14045.22
A14045.06EA 172 EUR
A14045.06 A14045.14 A14045.22
Tropyliumtetrafluoroborat ≥97%
Tropyliumtetrafluoroborat

Purity: 97% CAS No.: 27081-10-3 Molecular Formula: C7H7BF4 Formula Weight: 177.94 Storage Temperature: 2°C to 8°C Melting Point: ca 240° dec. Beilstein Reference: 3574512 UN No.: 1759 MDL No.: MFCD00013402

Formel: C₇H₇BF₄
Molecular Weight: 177,94 g/mol
Schmelzpunkt: ∼ 240 °C (Zers.)
MDL: MFCD00013402
CAS-Nummer: 27081-10-3
EINECS: 248-214-5
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR