Tropyliumtetrafluoroborat

Lieferant: CHEMPUR

Synonyme: Tropyliumtetrafluoroborate, Cyclohepta-2,4,6-trienylium tetrafluoroborate

FL9408.POR 009408-25G
CHMPFL9408.POREA 0 EUR
CHMPFL9408.POR FLUO009408-25G
Tropyliumtetrafluoroborat
Tropyliumtetrafluoroborat
Formel: C₇H₇BF₄
Molecular Weight: 177,94 g/mol
MDL: MFCD00013402
CAS-Nummer: 27081-10-3
EINECS: 248-214-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR