2-(4-Bromphenyl)-6-methylimidazo[1,2-a]pyridin
Lieferant: COMBI-BLOCKS
HC-2062-5G
HC-2062-1G
COBBHC-2062-5GEA
503
EUR
COBBHC-2062-5G
COBBHC-2062-1G
2-(4-Bromphenyl)-6-methylimidazo[1,2-a]pyridin
2-(4-Bromphenyl)-6-methylimidazo[1,2-a]pyridin
Formel:
C₁₄H₁₁BrN₂ Molecular Weight: 287,16 g/mol |
MDL:
MFCD00444784 CAS-Nummer: 1023-01-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...