1-Methyl-1H-indazol-4-boronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole
PN-9017-1G
PN-9017-5G
COBBPN-9017-1GEA
218
EUR
COBBPN-9017-1G
COBBPN-9017-5G
1-Methyl-1H-indazol-4-boronsäurepinakolester
1-Methyl-1H-indazol-4-boronsäurepinakolester
Formel:
C₁₄H₁₉BN₂O₂ Molecular Weight: 258.1239 g/mol |
MDL:
MFCD10700158 CAS-Nummer: 885698-94-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...