2-Methyl-2H-indazol-5-boronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-indazole
PN-9132-5G
PN-9132-1G
PN-9132-250M
COBBPN-9132-5GEA
1100
EUR
COBBPN-9132-5G
COBBPN-9132-1G
COBBPN-9132-250M
2-Methyl-2H-indazol-5-boronsäurepinakolester
2-Methyl-2H-indazol-5-boronsäurepinakolester
Formel:
C₁₄H₁₉BN₂O₂ Molecular Weight: 258.1239 g/mol |
MDL:
MFCD11109436 CAS-Nummer: 1189746-27-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...