1-(Methoxymethyl)-1H-indazol-6-boronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
1-(Methoxymethyl)-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole
PN-6380-1G
PN-6380-250M
COBBPN-6380-1GEA
1110
EUR
COBBPN-6380-1G
COBBPN-6380-250M
1-(Methoxymethyl)-1H-indazol-6-boronsäurepinakolester
1-(Methoxymethyl)-1H-indazol-6-boronsäurepinakolester
Formel:
C₁₅H₂₁BN₂O₃ Molecular Weight: 288.1498 g/mol |
MDL:
MFCD16295119 CAS-Nummer: 1256360-14-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...