2-Methyl-2H-indazol-4-boronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Methyl-2H-indazol-4-yl)boronic acid

BB-9026-1G BB-9026-250M
COBBBB-9026-1GEA 1250 EUR
COBBBB-9026-1G COBBBB-9026-250M
2-Methyl-2H-indazol-4-boronsäure
2-Methyl-2H-indazole-4-boronic acid
Formel: C₈H₉BN₂O₂
Molecular Weight: 175,98 g/mol
MDL: MFCD09870055
CAS-Nummer: 1001907-56-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR