5-Brom-2-methyl-2H-indazol

Lieferant: COMBI-BLOCKS

Synonyme: 2-Methyl-5-bromo-2H-indazole

IN-0204-5G IN-0204-1G
COBBIN-0204-5GEA 377 EUR
COBBIN-0204-5G COBBIN-0204-1G
5-Brom-2-methyl-2H-indazol
5-Bromo-2-methyl-2H-indazole
Formel: C₈H₇BrN₂
Molecular Weight: 211,0600 g/mol
MDL: MFCD09870047
CAS-Nummer: 465529-56-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR