3-Amino-5-brom-1-methyl-1H-indazol 97%

Lieferant: Thermo Fisher Scientific
Achtung

H33202.MD H33202.06 H33202.03
H33202.MDEA 40.7 EUR
H33202.MD H33202.06 H33202.03
3-Amino-5-brom-1-methyl-1H-indazol 97%
3-Amino-5-bromo-1-methyl-1H-indazole
Formel: C₈H₈BrN₃
Molecular Weight: 226.08 g/mol
Schmelzpunkt: 133…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09864804
CAS-Nummer: 1000018-06-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR