1-Acetyl-5-nitro-1H-indazol

Lieferant: COMBI-BLOCKS

Synonyme: 1-(5-Nitro-1H-indazol-1-yl)ethanone

HC-6242-5G HC-6242-1G
COBBHC-6242-5GEA 404 EUR
COBBHC-6242-5G COBBHC-6242-1G
1-Acetyl-5-nitro-1H-indazol
1-Acetyl-5-nitro-1H-indazol
Formel: C₉H₇N₃O₃
Molecular Weight: 205,17 g/mol
MDL: MFCD09261292
CAS-Nummer: 13436-55-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR