3-Amino-5-(trifluormethyl)-1H-indazol
Lieferant: BIONET RESEARCH
Synonyme:
5-(Trifluoromethyl)-1H-indazol-3-amine
10Y-0702.1
10Y-0702.10
10Y-0702.5
10Y-0702.500
BIOT10Y-0702.1EA
0
EUR
BIOT10Y-0702.1
BIOT10Y-0702.10
BIOT10Y-0702.5
BIOT10Y-0702.500
3-Amino-5-(trifluormethyl)-1H-indazol
3-Amino-5-(trifluoromethyl)-1H-indazole
Formel:
C₈H₆F₃N₃ Molecular Weight: 201,15 g/mol |
MDL:
MFCD01719740 CAS-Nummer: 2250-53-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...