2-Methyl-2H-indazol

Lieferant: COMBI-BLOCKS

HI-1553-5G HI-1553-1G
COBBHI-1553-5GEA 1020 EUR
COBBHI-1553-5G COBBHI-1553-1G
2-Methyl-2H-indazol
2-Methyl-2H-indazole
Formel: C₈H₈N₂
Molecular Weight: 132,162 g/mol
MDL: MFCD00234418
CAS-Nummer: 4838-00-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR