Bis(pyridin)iodoniumtetrafluoroborat 97%

Lieferant: Thermo Fisher Scientific
Achtung

369680010 369680050
ACRO369680010EA 70.8 EUR
ACRO369680010 ACRO369680050
Bis(pyridin)iodoniumtetrafluoroborat 97%
Bis(pyridin)iodoniumtetrafluoroborat
Formel: C₁₀H₁₀BF₄IN₂
Molecular Weight: 371.91 g/mol
Schmelzpunkt: 137…141 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03703393
CAS-Nummer: 15656-28-7

Bestellen

Spezifikation Testergebnisse

Appearance Light yellow to orange to tan Powder or crystals
Infrared spectrum Conforms
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR