[3-Methyl-5-phenyl-4-isoxazolyl]methanol

Lieferant: Thermo Scientific

CC20309.250 CC20309.1
MAYBCC20309.250EA 0 EUR
MAYBCC20309.250 MAYBCC20309.1
[3-Methyl-5-phenyl-4-isoxazolyl]methanol
[3-Methyl-5-phenyl-4-isoxazolyl]methanol
Formel: C₁₁H₁₁NO₂
CAS-Nummer: 113826-87-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR