3-(Chlormethyl)-1,2,4-oxadiazol

Lieferant: BIONET RESEARCH

11X-0842.1 11X-0842.10 11X-0842.5 11X-0842.500
BIOT11X-0842.1EA 0 EUR
BIOT11X-0842.1 BIOT11X-0842.10 BIOT11X-0842.5 BIOT11X-0842.500
3-(Chlormethyl)-1,2,4-oxadiazol
3-(Chloromethyl)-1,2,4-oxadiazole
Formel: C₃H₃ClN₂O
Molecular Weight: 118.521 g/mol
MDL: MFCD00052097
CAS-Nummer: 51791-12-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR