2-(Chlormethyl)oxazol

Lieferant: CHEMPUR

56-0003.1 56-0003.5
CHMP56-0003.1EA 0 EUR
CHMP56-0003.1 CHMP56-0003.5
2-(Chlormethyl)oxazol
2-(Chloromethyl)oxazole
Formel: C₄H₄ClNO
Molecular Weight: 117,53 g/mol
MDL: MFCD04114926
CAS-Nummer: 185246-17-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR