Methyl-5-methyl-4-oxazolcarboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 5-methyloxazole-4-carboxylate

HI-1247-5G HI-1247-1G
COBBHI-1247-5GEA 1010 EUR
COBBHI-1247-5G COBBHI-1247-1G
Methyl-5-methyl-4-oxazolcarboxylat
Methyl-5-methyl-4-oxazolcarboxylat
Formel: C₆H₇NO₃
Molecular Weight: 141.1247 g/mol
MDL: MFCD08689502
CAS-Nummer: 41172-57-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR