Ethyl-2-amino-4-methyl-5-oxazolcarboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 2-amino-4-methyloxazole-5-carboxylate

HC-2030-1G HC-2030-250M
COBBHC-2030-1GEA 462 EUR
COBBHC-2030-1G COBBHC-2030-250M
Ethyl-2-amino-4-methyl-5-oxazolcarboxylat
Ethyl-2-amino-4-methyl-5-oxazolcarboxylat
Formel: C₇H₁₀N₂O₃
Molecular Weight: 170.1659 g/mol
MDL: MFCD09028430
CAS-Nummer: 79221-15-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR