2-Amino-4-methyloxazol

Lieferant: FLUOROCHEM

Synonyme: 4-Methyloxazol-2-amine

016110-1G 016110-5G
FLUO016110-1GEA 128 EUR
FLUO016110-1G FLUO016110-5G
2-Amino-4-methyloxazol
2-Amino-4-methyloxazole
Formel: C₄H₆N₂O
Molecular Weight: 98.1032 g/mol
MDL: MFCD00126682
CAS-Nummer: 35629-70-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR