2-Amino-4-methyloxazol

Lieferant: Thermo Scientific

Synonyme: 4-Methyloxazol-2-amine

KM08202DA KM08202EA
MAYBKM08202DAEA 0 EUR
MAYBKM08202DA MAYBKM08202EA
2-Amino-4-methyloxazol
2-Amino-4-methyloxazole
Formel: C₄H₆N₂O
Molecular Weight: 98,1032 g/mol
MDL: MFCD00126682
CAS-Nummer: 35629-70-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR