Oxazol-5-methanol

Lieferant: COMBI-BLOCKS

Synonyme: Oxazol-5-ylmethanol, 5-(Hydroxymethyl)-1,3-oxazole

HI-1270-1G HI-1270-5G
COBBHI-1270-1GEA 385 EUR
COBBHI-1270-1G COBBHI-1270-5G
Oxazol-5-methanol
Oxazol-5-methanol
Formel: C₄H₅NO₂
Molecular Weight: 99,0880 g/mol
MDL: MFCD09027272
CAS-Nummer: 127232-41-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR