Okadasäure ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
9,10-deepithio-9,10-didehydroacanthifolicin, OA
J60155.XS
J60155.XG
J60155.IX
J60155.XSEA
438
EUR
J60155.XS
J60155.XG
J60155.IX
Okadasäure ≥98%
Okadaic acid
Formel:
C₄₄H₆₈O₁₃ Molecular Weight: 805,02 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00083455 CAS-Nummer: 78111-17-8 UN: 3462 ADR: 6.1,I Merck Index: 14,6819 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...