Okadasäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 9,10-deepithio-9,10-didehydroacanthifolicin, OA

M-Clarity™ Qualitätsstufe = MQ200

O9381-.1MG O9381-25UG
SIALO9381-.1MGEA 877 EUR
SIALO9381-.1MG SIALO9381-25UG
Okadasäure, Sigma-Aldrich®
Okadaic acid
Formel: C₄₄H₆₈O₁₃
Molecular Weight: 805,02 g/mol
MDL: MFCD00083455
CAS-Nummer: 78111-17-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR