Okadasäure Kaliumsalz 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Halochondrine A potassium salt, OA potassium salt, 9,10-deepithio-9,10-didehydroacanthifolicin potassium salt
J61791.XS
J61791.XG
J61791.IX
J61791.XSEA
312
EUR
J61791.XS
J61791.XG
J61791.IX
Okadasäure Kaliumsalz 98%
Okadaic acid potassium salt
Potent inhibitor of serine/threonine-specific protein phosphatases 1 and 2A (PP1 and PP2A). Does not inhibit tyrosine or alkaline phosphatases
Formel:
C₄₄H₆₇O₁₃·K Molecular Weight: 843.11 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00214356 CAS-Nummer: 155751-72-7 UN: 3462 ADR: 6.1 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...