Okadasäure Kaliumsalz, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ200
SIALO7885-10UGEA
82.1
EUR
SIALO7885-10UG
SIALO7885-25UG
SIALO7885-50UG
Okadasäure Kaliumsalz, Sigma-Aldrich®
Okadaic acid potassium salt
Formel:
C₄₄H₆₇O₁₃·K Molecular Weight: 843,11 g/mol |
MDL:
MFCD00214356 CAS-Nummer: 155751-72-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...