Ethylpyrazolo[1,5-a]pyridin-2-carboxylat

Lieferant: CHEMPUR

68-0050.500 68-0050.1
CHMP68-0050.500EA 0 EUR
CHMP68-0050.500 CHMP68-0050.1
Ethylpyrazolo[1,5-a]pyridin-2-carboxylat
Ethylpyrazolo[1,5-a]pyridin-2-carboxylat
Formel: C₁₀H₁₀N₂O₂
Molecular Weight: 190.1986 g/mol
MDL: MFCD04115107
CAS-Nummer: 80537-14-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR