2-(6-Chlor-3-pyridazinyl)-2-(2,4-dichlorphenyl)acetonitril

Lieferant: BIONET RESEARCH

6D-001.500 6D-001.1 6D-001.5 6D-001.10
BIOT6D-001.500EA 0 EUR
BIOT6D-001.500 BIOT6D-001.1 BIOT6D-001.5 BIOT6D-001.10
2-(6-Chlor-3-pyridazinyl)-2-(2,4-dichlorphenyl)acetonitril
2-(6-Chlor-3-pyridazinyl)-2-(2,4-dichlorphenyl)acetonitril
Formel: C₁₂H₆Cl₃N₃
MDL: MFCD00140702
CAS-Nummer: 209412-09-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR