2-(6-Chlor-3-pyridazinyl)-2-(2,6-dichlorphenyl)acetonitril
Lieferant: BIONET RESEARCH
1F-066.500
1F-066.1
1F-066.5
BIOT1F-066.500EA
0
EUR
BIOT1F-066.500
BIOT1F-066.1
BIOT1F-066.5
2-(6-Chlor-3-pyridazinyl)-2-(2,6-dichlorphenyl)acetonitril
2-(6-Chlor-3-pyridazinyl)-2-(2,6-dichlorphenyl)acetonitril
Formel:
C₁₂H₆Cl₃N₃ |
MDL:
MFCD00202119 CAS-Nummer: 209412-06-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...